Assay Details
Target Gene Details
Entrez Gene ID: | 6830 |
Gene Name: | SPT6 homolog, histone chaperone |
Gene Aliases: |
SPT6, SPT6H, emb-5 |
Location: |
Chr.17:28662183-28702684 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUPT6H | NM_001320755.1 | NP_001307684.1 | ||
NM_003170.4 | NP_003161.2 | |||
XM_017024978.1 | XP_016880467.1 | |||
AL833607.1 | ||||
BC136522.1 | ||||
BC136524.1 | ||||
BC150268.1 | ||||
D79984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain | PROCA1 SUPT6H TLCD1 TRAF4 PIGS RPL23A NARR RPS7P1 ALDOC SNORD42A KIAA0100 SNORD42B RAB34 FOXN1 SPAG5 SNORD4B SGK494 SNORD4A SDF2 UNC119 NEK8 SPAG5-AS1 SLC13A2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |