Assay Details
Target Gene Details
Entrez Gene ID: | 5578 |
Gene Name: | protein kinase C alpha |
Gene Aliases: |
AAG6, PKC-alpha, PKCA, PRKACA |
Location: |
Chr.17:66302640-66810744 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKCA | NM_002737.2 | NP_002728.1 | ||
XM_017024836.1 | XP_016880325.1 | |||
XM_017024841.1 | XP_016880330.1 | |||
AB209475.1 | BAD92712.1 | |||
AB451258.1 | ||||
AB451383.1 | ||||
AY633609.1 | AAV33302.1 | |||
BC053321.1 | AAH53321.1 | |||
BC109273.1 | AAI09274.1 | |||
BC109274.1 | AAI09275.1 | |||
LC151409.1 | ||||
M22199.1 | AAA60098.1 | |||
X52479.1 | CAA36718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510721 | Chr.17:66240899 - 66313640 on Build GRCh38 | Deletion | PRKCA |
nsv526542 | Chr.17:66292519 - 66307675 on Build GRCh38 | Loss | PRKCA |
nsv833520 | Chr.17:66261176 - 66438257 on Build GRCh38 | Gain | PRKCA-AS1 PRKCA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |