Assay Details
Target Gene Details
Entrez Gene ID: | 163 |
Gene Name: | adaptor related protein complex 2 beta 1 subunit |
Gene Aliases: |
ADTB2, AP105B, AP2-BETA, CLAPB1 |
Location: |
Chr.17:35586899-35726417 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2038 | Chr.17:35569617 - 35598796 on Build GRCh38 | Insertion | LOC105371743 PEX12 SNORD7 AP2B1 |
esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss | RASL10B SLFN12L LRRC37A8P C17orf50 SLFN11 GAS2L2 RDM1 SLFN12 CCL5 MMP28 TAF15 AP2B1 SLFN5 SLFN14 HEATR9 LOC105371933 LOC105371743 SLFN13 PEX12 SNORD7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |