Assay Details
Target Gene Details
Entrez Gene ID: | 10642 |
Gene Name: | insulin like growth factor 2 mRNA binding protein 1 |
Gene Aliases: |
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1 |
Location: |
Chr.17:48997412-49056145 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGF2BP1 | NM_001160423.1 | NP_001153895.1 | ||
NM_006546.3 | NP_006537.3 | |||
XM_011524201.2 | XP_011522503.1 | |||
XM_017024022.1 | XP_016879511.1 | |||
AF117106.1 | AAD09826.1 | |||
AF198254.1 | AAF37203.1 | |||
AK313229.1 | ||||
BP238821.1 | ||||
DQ227344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain | IGF2BP1 UBE2Z GIP SNF8 LOC105371814 ATP5G1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |