Assay Details
Target Gene Details
Entrez Gene ID: | 15 |
Gene Name: | aralkylamine N-acetyltransferase |
Gene Aliases: |
DSPS, SNAT |
Location: |
Chr.17:76453351-76470797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AANAT | NM_001166579.1 | NP_001160051.1 | ||
BC092430.1 | AAH92430.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833549 | Chr.17:76365444 - 76527890 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)