Assay Details
Target Gene Details
Entrez Gene ID: | 101928418 |
Gene Name: | uncharacterized LOC101928418 |
Gene Aliases: |
- |
Location: |
Chr.17:12671862-12706135 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928418 | NR_104605.1 |
Target Gene Details
Entrez Gene ID: | 93649 |
Gene Name: | myocardin |
Gene Aliases: |
MYCD |
Location: |
Chr.17:12665890-12768949 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYOCD | NM_001146312.2 | NP_001139784.1 | ||
NM_153604.3 | NP_705832.1 | |||
XM_005256863.1 | XP_005256920.1 | |||
XM_005256864.1 | XP_005256921.1 | |||
XM_017025342.1 | XP_016880831.1 | |||
AF532596.1 | AAN33040.1 | |||
AK128608.1 | ||||
AK292885.1 | ||||
AY764180.1 | AAV33439.1 | |||
BC126307.1 | ||||
BC143391.1 | ||||
DA650534.1 | ||||
DB226467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34095 | Chr.17:12436261 - 12841981 on Build GRCh38 | Loss | MYOCD LOC101928418 LOC100128006 ARHGAP44 LINC00670 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |