Assay Details
Target Gene Details
Entrez Gene ID: | 79777 |
Gene Name: | acyl-CoA binding domain containing 4 |
Gene Aliases: |
HMFT0700 |
Location: |
Chr.17:45132600-45144181 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953903 | Chr.17:45081533 - 45148633 on Build GRCh38 | Deletion | NMT1 HEXIM1 PLCD3 MIR6784 ACBD4 |
nsv527788 | Chr.17:44899824 - 45226241 on Build GRCh38 | Loss | DCAKD MIR6784 ACBD4 C1QL1 HEXIM2 MIR6783 GFAP LOC107987243 KIF18B NMT1 HEXIM1 PLCD3 LOC105371795 CCDC103 FMNL1 LOC339192 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |