Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | GPR139 GPRC5B ERI2 LOC81691 CRYM-AS1 ANKS4B C16orf62 SYT17 ACSM1 LYRM1 CLEC19A ACSM3 DNAH3 UMOD ACSM5 CCP110 IQCK GP2 LOC102723385 GDE1 KNOP1 TMC7 NPIPB3 ACSM2B SMG1P3 PDILT TMEM159 DCUN1D3 THUMPD1 TMC5 ZP2 SNX29P1 LOC100190986 CRYM ACSM2A ITPRIPL2 COQ7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |