Assay Details
Target Gene Details
Entrez Gene ID: | 85320 |
Gene Name: | ATP binding cassette subfamily C member 11 |
Gene Aliases: |
EWWD, MRP8, WW |
Location: |
Chr.16:48164842-48249973 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCC11 | XM_017023795.1 | XP_016879284.1 | ||
XM_017023796.1 | XP_016879285.1 | |||
XM_017023797.1 | XP_016879286.1 | |||
XM_017023798.1 | XP_016879287.1 | |||
XM_017023799.1 | XP_016879288.1 | |||
XM_017023800.1 | XP_016879289.1 | |||
XM_017023801.1 | XP_016879290.1 | |||
XM_017023803.1 | XP_016879292.1 | |||
AF352582.1 | AAK19755.1 | |||
AK301547.1 |
Target Gene Details
Entrez Gene ID: | 83752 |
Gene Name: | lon peptidase 2, peroxisomal |
Gene Aliases: |
LONP, LONPL |
Location: |
Chr.16:48244167-48353979 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LONP2 | NM_001300948.1 | NP_001287877.1 | ||
NM_031490.3 | NP_113678.2 | |||
DC393124.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |