Assay Details
Target Gene Details
Entrez Gene ID: | 5579 |
Gene Name: | protein kinase C beta |
Gene Aliases: |
PKC-beta, PKCB, PRKCB1, PRKCB2 |
Location: |
Chr.16:23835979-24220611 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKCB | NM_002738.6 | NP_002729.2 | ||
NM_212535.2 | NP_997700.1 | |||
BC036472.1 | AAH36472.1 | |||
BI549165.1 | ||||
DB456157.1 | ||||
M13975.1 | AAA60095.1 | |||
X06318.1 | CAA29634.1 | |||
X07109.1 | CAA30130.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)