Assay Details
Target Gene Details
Entrez Gene ID: | 204474 |
Gene Name: | protein disulfide isomerase like, testis expressed |
Gene Aliases: |
PDIA7 |
Location: |
Chr.16:20359170-20404742 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDILT | NM_174924.1 | NP_777584.1 | ||
XM_011545764.1 | XP_011544066.1 | |||
XM_011545765.1 | XP_011544067.1 | |||
XM_011545766.2 | XP_011544068.1 | |||
AK097476.1 | BAC05068.1 | |||
BC042607.1 | AAH42607.1 | |||
BC044936.1 | AAH44936.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047399 | Chr.16:20378305 - 20522307 on Build GRCh38 | Gain | ACSM5 PDILT ACSM2A |
dgv2795n100 | Chr.16:20325210 - 20411678 on Build GRCh38 | Gain | UMOD GP2 ACSM5 PDILT |
dgv4942n54 | Chr.16:20340615 - 20466152 on Build GRCh38 | Gain | UMOD ACSM5 PDILT ACSM2A |
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | SYT17 NPIPB3 SMG1P3 TMC5 DCUN1D3 GDE1 LOC100190986 CLEC19A ANKS4B LYRM1 ACSM2B GPRC5B GPR139 COQ7 ACSM2A ACSM1 IQCK LOC81691 C16orf62 THUMPD1 CCP110 CRYM SNX29P1 ACSM5 ZP2 KNOP1 ERI2 TMEM159 TMC7 UMOD ACSM3 GP2 ITPRIPL2 CRYM-AS1 PDILT LOC102723385 DNAH3 |
esv3638138 | Chr.16:20348867 - 20489664 on Build GRCh38 | Gain | UMOD ACSM5 PDILT ACSM2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |