Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | IQCK GP2 PDILT KNOP1 SMG1P3 ACSM5 TMC5 C16orf62 ACSM3 ACSM2A TMC7 LOC100190986 GPRC5B ANKS4B ERI2 GDE1 DCUN1D3 LOC81691 ITPRIPL2 GPR139 THUMPD1 CLEC19A NPIPB3 ZP2 LOC102723385 ACSM1 ACSM2B DNAH3 LYRM1 TMEM159 CRYM SNX29P1 SYT17 UMOD CRYM-AS1 COQ7 CCP110 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |