Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | CRYM CLEC19A ANKS4B ITPRIPL2 ACSM2A DCUN1D3 THUMPD1 ACSM1 CCP110 C16orf62 UMOD NPIPB3 LOC81691 TMC5 GP2 SMG1P3 ACSM5 COQ7 TMEM159 ACSM2B GPR139 LOC102723385 GDE1 IQCK ZP2 ERI2 LOC100190986 PDILT TMC7 LYRM1 SYT17 CRYM-AS1 KNOP1 SNX29P1 GPRC5B ACSM3 DNAH3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |