Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | ACSM2B DNAH3 SMG1P3 ERI2 SYT17 CRYM-AS1 DCUN1D3 COQ7 NPIPB3 ACSM3 ANKS4B GPRC5B GDE1 CLEC19A IQCK ZP2 LOC100190986 KNOP1 ACSM5 GPR139 ACSM2A TMC7 THUMPD1 TMC5 GP2 ITPRIPL2 UMOD SNX29P1 LOC81691 LYRM1 CRYM ACSM1 C16orf62 TMEM159 PDILT LOC102723385 CCP110 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |