Assay Details
Target Gene Details
Entrez Gene ID: | 164 |
Gene Name: | adaptor related protein complex 1 gamma 1 subunit |
Gene Aliases: |
ADTG, CLAPG1 |
Location: |
Chr.16:71729000-71809073 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP1G1 | NM_001030007.1 | NP_001025178.1 | ||
NM_001128.5 | NP_001119.3 | |||
AB015317.1 | BAA33389.1 | |||
AK055122.1 | ||||
AK128078.1 | ||||
AK225906.1 | ||||
AK293233.1 | ||||
AK314484.1 | ||||
AL110198.1 | CAB53673.1 | |||
BC036283.1 | AAH36283.1 | |||
Y12226.1 | CAA72902.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |