Assay Details
Target Gene Details
Entrez Gene ID: | 221184 |
Gene Name: | copine 2 |
Gene Aliases: |
COPN2, CPN2 |
Location: |
Chr.16:57092543-57147966 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPNE2 | NM_152727.5 | NP_689940.3 | ||
AF492484.1 | AAO85483.1 | |||
AK315394.1 | ||||
BC053658.1 | AAH53658.1 | |||
BG830959.1 | ||||
BG831872.1 | ||||
DB504175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv827683 | Chr.16:57016822 - 57147283 on Build GRCh38 | Loss | CPNE2 NLRC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |