Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | THUMPD1 ACSM3 SNX29P1 ACSM2A ACSM5 GPRC5B LOC102723385 LOC100190986 ITPRIPL2 ANKS4B TMC5 NPIPB3 UMOD SYT17 ACSM2B ERI2 LYRM1 GDE1 LOC81691 CCP110 CRYM-AS1 TMC7 COQ7 ZP2 IQCK GP2 CLEC19A ACSM1 PDILT CRYM DCUN1D3 KNOP1 GPR139 SMG1P3 C16orf62 TMEM159 DNAH3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |