Assay Details
Target Gene Details
Entrez Gene ID: | 80205 |
Gene Name: | chromodomain helicase DNA binding protein 9 |
Gene Aliases: |
AD013, CHD-9, CReMM, KISH2, PRIC320 |
Location: |
Chr.16:53054991-53327502 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHD9 | NM_025134.4 | NP_079410.4 | ||
XM_005256168.3 | XP_005256225.1 | |||
XM_017023719.1 | XP_016879208.1 | |||
XM_017023720.1 | XP_016879209.1 | |||
AY647157.1 | AAT66509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066719 | Chr.16:52261069 - 53107993 on Build GRCh38 | Loss | CASC16 LOC105371267 CHD9 TOX3 CASC22 |
nsv833232 | Chr.16:52974938 - 53157922 on Build GRCh38 | Loss | LOC105371267 CHD9 |
nsv833233 | Chr.16:53046019 - 53257336 on Build GRCh38 | Gain | LOC105371267 CHD9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |