Assay Details
Target Gene Details
Entrez Gene ID: | 1006 |
Gene Name: | cadherin 8 |
Gene Aliases: |
Nbla04261 |
Location: |
Chr.16:61640435-62036835 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH8 | NM_001796.4 | NP_001787.2 | ||
XM_005255760.3 | XP_005255817.1 | |||
XM_017022829.1 | XP_016878318.1 | |||
AB035305.1 | BAA87419.1 | |||
AK124734.1 | ||||
BC113416.1 | ||||
L34060.1 | AAA35628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833256 | Chr.16:61558794 - 61736777 on Build GRCh38 | Gain | CDH8 |
nsv572844 | Chr.16:61625455 - 61690440 on Build GRCh38 | Loss | CDH8 |
esv2762088 | Chr.16:61220267 - 61799347 on Build GRCh38 | Gain+Loss | CDH8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |