Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | IQCK LOC102723385 DCUN1D3 CRYM-AS1 TMC7 ZP2 GPRC5B ACSM3 ACSM2B CCP110 NPIPB3 LOC100190986 PDILT UMOD SNX29P1 CLEC19A LYRM1 C16orf62 LOC81691 ACSM1 TMEM159 GDE1 SYT17 KNOP1 THUMPD1 ACSM2A ITPRIPL2 COQ7 GP2 TMC5 ANKS4B ACSM5 GPR139 ERI2 SMG1P3 DNAH3 CRYM |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |