Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | SNX29P1 ERI2 ANKS4B SMG1P3 DCUN1D3 NPIPB3 KNOP1 THUMPD1 GP2 ZP2 ACSM5 ACSM1 LOC100190986 ACSM2A ACSM2B CLEC19A COQ7 SYT17 TMC5 UMOD LYRM1 TMEM159 ITPRIPL2 LOC102723385 GDE1 IQCK TMC7 GPR139 CRYM-AS1 GPRC5B CCP110 DNAH3 LOC81691 ACSM3 PDILT CRYM C16orf62 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |