Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | THUMPD1 ACSM5 COQ7 PDILT IQCK CRYM-AS1 LOC81691 C16orf62 NPIPB3 CRYM LOC100190986 GDE1 ANKS4B ITPRIPL2 GPRC5B UMOD GPR139 SNX29P1 SYT17 ACSM2B CCP110 ERI2 DNAH3 ZP2 TMC7 KNOP1 LOC102723385 SMG1P3 ACSM3 CLEC19A ACSM1 LYRM1 GP2 TMEM159 DCUN1D3 ACSM2A TMC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |