Assay Details
Target Gene Details
Entrez Gene ID: | 1540 |
Gene Name: | CYLD lysine 63 deubiquitinase |
Gene Aliases: |
BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2 |
Location: |
Chr.16:50742026-50801935 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYLD | NM_001042355.1 | NP_001035814.1 | ||
NM_001042412.1 | NP_001035877.1 | |||
NM_015247.2 | NP_056062.1 | |||
XM_005255812.2 | XP_005255869.1 | |||
XM_006721149.1 | XP_006721212.1 | |||
XM_011522907.2 | XP_011521209.1 | |||
XM_011522908.1 | XP_011521210.1 | |||
XM_011522911.2 | XP_011521213.1 | |||
XM_017022977.1 | XP_016878466.1 | |||
XM_017022978.1 | XP_016878467.1 | |||
XM_017022979.1 | XP_016878468.1 | |||
XM_017022980.1 | XP_016878469.1 | |||
XM_017022981.1 | XP_016878470.1 | |||
AF161542.1 | AAF29029.1 | |||
AJ250014.1 | CAB93533.1 | |||
AK056226.1 | ||||
AK225202.1 | ||||
AK292975.1 | ||||
AK310832.1 | ||||
BC012342.1 | AAH12342.1 | |||
BI458962.1 | ||||
DA785459.1 | ||||
DB119480.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952674 | Chr.16:50743190 - 50793689 on Build GRCh38 | Duplication | CYLD |
esv3638608 | Chr.16:50654254 - 50854693 on Build GRCh38 | Gain | LOC101927272 SNX20 CYLD MIR3181 NOD2 |
dgv2977n100 | Chr.16:50744296 - 50798809 on Build GRCh38 | Loss | CYLD |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |