Assay Details
Target Gene Details
Entrez Gene ID: | 54496 |
Gene Name: | protein arginine methyltransferase 7 |
Gene Aliases: |
- |
Location: |
Chr.16:68310974-68358832 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510691 | Chr.16:68266527 - 68325431 on Build GRCh38 | Deletion | SLC7A6OS SLC7A6 PRMT7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |