Assay Details
Target Gene Details
Entrez Gene ID: | 766 |
Gene Name: | carbonic anhydrase 7 |
Gene Aliases: |
CAVII |
Location: |
Chr.16:66844379-66854149 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CA7 | NM_001014435.1 | NP_001014435.1 | ||
NM_005182.2 | NP_005173.1 | |||
XM_005256136.3 | XP_005256193.1 | |||
XM_011523311.1 | XP_011521613.1 | |||
XM_011523312.1 | XP_011521614.1 | |||
AY075019.1 | AAL78167.1 | |||
AY075020.1 | AAL78168.1 | |||
BC033865.1 | AAH33865.1 | |||
BI752037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510690 | Chr.16:66817285 - 66858281 on Build GRCh38 | Deletion | CA7 NAE1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |