Assay Details
Target Gene Details
Entrez Gene ID: | 84706 |
Gene Name: | glutamic--pyruvic transaminase 2 |
Gene Aliases: |
ALT2, GPT 2, MRT49 |
Location: |
Chr.16:46884380-46931297 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPT2 | NM_001142466.2 | NP_001135938.1 | ||
NM_133443.3 | NP_597700.1 | |||
XM_017023790.1 | XP_016879279.1 | |||
AK094971.1 | BAC04465.1 | |||
AY029173.2 | AAK31794.2 | |||
BC062555.1 | AAH62555.1 | |||
DA400677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1063660 | Chr.16:46896711 - 46944880 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)