Assay Details
Target Gene Details
Entrez Gene ID: | 100529251 |
Gene Name: | CKLF-CMTM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:66552563-66579135 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CKLF-CMTM1 | NM_001202509.1 | NP_001189438.1 | ||
NM_001204098.1 | NP_001191027.1 | |||
NM_001204099.1 | NP_001191028.1 | |||
BM929895.1 | ||||
FY211334.1 | ||||
FY212695.1 |
Target Gene Details
Entrez Gene ID: | 113540 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 1 |
Gene Aliases: |
CKLFH, CKLFH1, CKLFSF1 |
Location: |
Chr.16:66566391-66579135 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv123n111 | Chr.16:66568129 - 66581930 on Build GRCh38 | Duplication | CKLF-CMTM1 CMTM1 CMTM2 |
esv3638871 | Chr.16:66570632 - 66577424 on Build GRCh38 | Gain | CKLF-CMTM1 CMTM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |