Assay Details
Target Gene Details
Entrez Gene ID: | 1428 |
Gene Name: | crystallin mu |
Gene Aliases: |
DFNA40, THBP |
Location: |
Chr.16:21258518-21303136 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYM | NM_001888.4 | NP_001879.1 | ||
AK290852.1 | ||||
AK310424.1 | ||||
BC018061.1 | AAH18061.1 | |||
BI860684.1 | ||||
BX648477.1 | CAI46030.1 | |||
L02950.1 | AAC16914.1 | |||
U85772.1 | AAB81564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638165 | Chr.16:21188383 - 21267495 on Build GRCh38 | Gain | CRYM ZP2 ANKS4B |
nsv571623 | Chr.16:21084049 - 21341186 on Build GRCh38 | Gain | CRYM CRYM-AS1 TMEM159 ZP2 ANKS4B DNAH3 |
esv3638164 | Chr.16:21185596 - 21338776 on Build GRCh38 | Gain | CRYM CRYM-AS1 ZP2 ANKS4B |
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | CRYM DCUN1D3 CRYM-AS1 KNOP1 GP2 THUMPD1 UMOD LOC102723385 TMC5 ACSM2A CLEC19A GDE1 PDILT GPR139 IQCK SMG1P3 DNAH3 TMC7 ACSM2B ERI2 LYRM1 LOC100190986 TMEM159 ANKS4B GPRC5B ACSM3 NPIPB3 ACSM1 CCP110 ACSM5 SYT17 SNX29P1 ZP2 COQ7 ITPRIPL2 C16orf62 LOC81691 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |