Assay Details
Target Gene Details
Entrez Gene ID: | 11245 |
Gene Name: | G protein-coupled receptor 176 |
Gene Aliases: |
HB-954 |
Location: |
Chr.15:39795049-39920933 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR176 | NM_001271854.1 | NP_001258783.1 | ||
NM_001271855.1 | NP_001258784.1 | |||
NM_007223.2 | NP_009154.1 | |||
XM_011521167.2 | XP_011519469.1 | |||
XM_011521168.2 | XP_011519470.1 | |||
XM_017021873.1 | XP_016877362.1 | |||
XM_017021874.1 | XP_016877363.1 | |||
XM_017021875.1 | XP_016877364.1 | |||
XM_017021876.1 | XP_016877365.1 | |||
XM_017021877.1 | XP_016877366.1 | |||
XM_017021878.1 | XP_016877367.1 | |||
AK302715.1 | ||||
BC067106.1 | AAH67106.1 | |||
D38449.1 | BAA07481.1 | |||
JQ308174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832983 | Chr.15:39806565 - 39956638 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3569465 | Chr.15:39178507 - 39912020 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv832979 | Chr.15:39749727 - 39926206 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)