Assay Details
Target Gene Details
Entrez Gene ID: | 9543 |
Gene Name: | immunoglobulin superfamily DCC subclass member 3 |
Gene Aliases: |
HsT18880, PUNC |
Location: |
Chr.15:65327127-65378040 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGDCC3 | NM_004884.3 | NP_004875.2 | ||
XM_011522241.1 | XP_011520543.1 | |||
XM_011522243.1 | XP_011520545.1 | |||
XM_011522244.1 | XP_011520546.1 | |||
XM_011522245.2 | XP_011520547.1 | |||
AK095529.1 | ||||
BC042054.1 | AAH42054.1 | |||
BC067107.1 | AAH67107.1 | |||
CB988291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1036360 | Chr.15:65333881 - 65390204 on Build GRCh38 | Gain | IGDCC3 IGDCC4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |