Assay Details
Target Gene Details
Entrez Gene ID: | 105371031 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.15:89399716-89507395 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371031 | XM_017022800.1 | XP_016878289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain | TICRR MIR9-3 POLG ABHD2 RLBP1 LOC105371031 MIR6766 LINC00928 LOC105370962 MIR9-3HG FANCI RHCG |
nsv516532 | Chr.15:89389564 - 89405705 on Build GRCh38 | Loss | LOC105371031 MIR9-3HG |
nsv833086 | Chr.15:89275748 - 89437378 on Build GRCh38 | Loss | MIR9-3 POLG LOC105371031 MIR6766 LOC105370962 MIR9-3HG FANCI |