Assay Details
Target Gene Details
Entrez Gene ID: | 9960 |
Gene Name: | ubiquitin specific peptidase 3 |
Gene Aliases: |
SIH003, UBP |
Location: |
Chr.15:63504511-63594640 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP3 | NM_001256702.1 | NP_001243631.1 | ||
NM_006537.3 | NP_006528.2 | |||
NR_046341.1 | ||||
NR_046342.1 | ||||
XM_017022763.1 | XP_016878252.1 | |||
XM_017022764.1 | XP_016878253.1 | |||
XM_017022765.1 | XP_016878254.1 | |||
XM_017022766.1 | XP_016878255.1 | |||
XM_017022767.1 | XP_016878256.1 | |||
XM_017022768.1 | XP_016878257.1 | |||
AF073344.1 | AAD42992.1 | |||
AK300948.1 | ||||
AK301236.1 | ||||
AK301563.1 | ||||
AK307810.1 | ||||
AK314223.1 | ||||
AY461579.1 | AAT37507.1 | |||
BC018113.2 | AAH18113.1 | |||
BC065300.1 | ||||
BC100029.1 | AAI00030.1 | |||
BC107137.1 | AAI07138.1 | |||
BC107138.1 | AAI07139.1 | |||
BT007269.1 | AAP35933.1 | |||
BX647760.1 | ||||
DA557812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951974 | Chr.15:63505902 - 63536501 on Build GRCh38 | Duplication |
![]() |
nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv1048029 | Chr.15:63414026 - 63565535 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)