Assay Details
Target Gene Details
Entrez Gene ID: | 10021 |
Gene Name: | hyperpolarization activated cyclic nucleotide gated potassium channel 4 |
Gene Aliases: |
SSS2 |
Location: |
Chr.15:73319859-73369264 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HCN4 | NM_005477.2 | NP_005468.1 | ||
XM_011521148.2 | XP_011519450.1 | |||
AJ132429.1 | CAB42604.1 | |||
AJ238850.1 | CAB52754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524049 | Chr.15:73320492 - 73387960 on Build GRCh38 | Loss | LOC107984728 HCN4 |
nsv569949 | Chr.15:73320036 - 73387960 on Build GRCh38 | Loss | LOC107984728 HCN4 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | REC114 CPLX3 FAM219B CYP1A1 LOC105376731 LOXL1-AS1 RPP25 SCAMP5 CLK3 LOC283731 SEMA7A STRA6 LOC732265 GOLGA6C NEO1 CYP1A2 BBS4 UBL7 GOLGA6A LOC107984728 SCAMP2 HCN4 ARID3B COX5A LOXL1 C15orf59-AS1 ISLR EDC3 MIR4513 ISLR2 TBC1D21 CYP11A1 HIGD2B MIR6882 ADPGK-AS1 LOC729739 C15orf39 LOC102723750 ULK3 PML UBL7-AS1 C15orf59 CCDC33 LMAN1L NPTN CSK PPCDC STOML1 NPTN-IT1 CD276 MPI LOC101929333 MIR6881 ADPGK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |