Assay Details
Target Gene Details
Entrez Gene ID: | 2958 |
Gene Name: | general transcription factor IIA subunit 2 |
Gene Aliases: |
HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS |
Location: |
Chr.15:59638062-59657541 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF2A2 | NM_001320929.1 | NP_001307858.1 | ||
NM_001320930.1 | NP_001307859.1 | |||
NM_004492.2 | NP_004483.1 | |||
AK312015.1 | ||||
BC000287.2 | AAH00287.1 | |||
BC001919.1 | AAH01919.1 | |||
BT007362.1 | AAP36026.1 | |||
CB156109.1 | ||||
CR542192.1 | CAG46989.1 | |||
HY002503.1 | ||||
HY036972.1 | ||||
U14193.1 | AAA64951.1 | |||
U21242.1 | AAB58247.1 | |||
X81713.1 | CAA57357.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain | MIR2116 GCNT3 FAM81A LDHAL6B FAM63B BNIP2 GTF2A2 RNF111 MYO1E CCNB2 SLTM |
esv3569653 | Chr.15:59636874 - 59670724 on Build GRCh38 | Gain | BNIP2 GTF2A2 |
esv3636646 | Chr.15:59635720 - 59677766 on Build GRCh38 | Gain | BNIP2 GTF2A2 |
nsv569605 | Chr.15:59563027 - 59664354 on Build GRCh38 | Loss | GCNT3 BNIP2 GTF2A2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |