Assay Details
Target Gene Details
Entrez Gene ID: | 8125 |
Gene Name: | acidic nuclear phosphoprotein 32 family member A |
Gene Aliases: |
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32 |
Location: |
Chr.15:68778535-68820922 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANP32A | NM_006305.3 | NP_006296.1 | ||
AF025684.1 | AAB91548.1 | |||
AK127498.1 | ||||
AK223280.1 | BAD97000.1 | |||
AK312703.1 | ||||
AY007110.1 | ||||
AY349171.1 | AAQ79832.1 | |||
BC000608.2 | AAH00608.1 | |||
BC007200.1 | AAH07200.1 | |||
BC125143.1 | ||||
BT007436.1 | AAP36104.1 | |||
U60823.1 | AAC50570.1 | |||
U73477.1 | AAB39706.1 | |||
X75090.1 | CAA52981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833046 | Chr.15:68639374 - 68841420 on Build GRCh38 | Loss | ANP32A-IT1 ANP32A MIR4312 CORO2B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |