Assay Details
Target Gene Details
Entrez Gene ID: | 3480 |
Gene Name: | insulin like growth factor 1 receptor |
Gene Aliases: |
CD221, IGFIR, IGFR, JTK13 |
Location: |
Chr.15:98648539-98964530 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGF1R | NM_000875.4 | NP_000866.1 | ||
NM_001291858.1 | NP_001278787.1 | |||
XM_017022139.1 | XP_016877628.1 | |||
AB425196.1 | ||||
BC113610.1 | ||||
BC113612.1 | ||||
BC143721.1 | ||||
DR007209.1 | ||||
EU826611.1 | ||||
EU826612.1 | ||||
X04434.1 | CAA28030.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |