Assay Details
Target Gene Details
Entrez Gene ID: | 60490 |
Gene Name: | phosphopantothenoylcysteine decarboxylase |
Gene Aliases: |
MDS018, PPC-DC, coaC |
Location: |
Chr.15:75023533-75060180 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPCDC | NM_001301101.1 | NP_001288030.1 | ||
NM_001301102.1 | NP_001288031.1 | |||
NM_001301103.1 | NP_001288032.1 | |||
NM_021823.4 | NP_068595.3 | |||
NR_125369.1 | ||||
XM_017022461.1 | XP_016877950.1 | |||
XM_017022464.1 | XP_016877953.1 | |||
XM_017022465.1 | XP_016877954.1 | |||
AK027491.1 | BAB55151.1 | |||
AY358848.1 | AAQ89207.1 | |||
BF243417.1 | ||||
DB488540.1 | ||||
DW437029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527237 | Chr.15:75012699 - 75028921 on Build GRCh38 | Loss | PPCDC SCAMP5 |
nsv569993 | Chr.15:74949814 - 75191411 on Build GRCh38 | Gain | PPCDC SCAMP5 RPP25 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | COX5A SCAMP5 LOXL1-AS1 CSK EDC3 NPTN-IT1 C15orf59 STRA6 GOLGA6A BBS4 LMAN1L MIR4513 CPLX3 CYP1A1 PML MIR6881 ISLR2 CD276 NEO1 ADPGK-AS1 LOC102723750 CYP1A2 CLK3 CCDC33 FAM219B GOLGA6C LOC101929333 ULK3 NPTN HIGD2B SEMA7A CYP11A1 C15orf39 LOC283731 RPP25 SCAMP2 MPI UBL7-AS1 C15orf59-AS1 LOC732265 HCN4 LOC729739 PPCDC TBC1D21 REC114 LOXL1 UBL7 MIR6882 ADPGK LOC105376731 STOML1 LOC107984728 ARID3B ISLR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |