Assay Details
Target Gene Details
Entrez Gene ID: | 54866 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 14D |
Gene Aliases: |
CPI17-like, GBPI-1, GBPI1 |
Location: |
Chr.15:40815445-40828793 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R14D | NM_001130143.1 | NP_001123615.1 | ||
NM_017726.7 | NP_060196.1 | |||
XM_017022372.1 | XP_016877861.1 | |||
AK000258.1 | BAA91037.1 | |||
AY050671.1 | AAL25829.1 | |||
BC096716.1 | AAH96716.1 | |||
BC098124.1 | AAH98124.1 | |||
BC098150.1 | AAH98150.1 | |||
BC098341.1 | AAH98341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054764 | Chr.15:40776427 - 40904212 on Build GRCh38 | Gain | DNAJC17 ZFYVE19 RHOV PPP1R14D SPINT1 VPS18 |
nsv569228 | Chr.15:40814287 - 40901953 on Build GRCh38 | Loss | ZFYVE19 RHOV PPP1R14D SPINT1 VPS18 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |