Assay Details
Target Gene Details
Entrez Gene ID: | 80765 |
Gene Name: | StAR related lipid transfer domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.15:81312666-81324183 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD5 | NM_181900.2 | NP_871629.1 | ||
NR_135013.1 | ||||
XM_017022639.1 | XP_016878128.1 | |||
AF480304.1 | AAL89654.1 | |||
AK026352.1 | ||||
AK097024.1 | ||||
AK225479.1 | ||||
AL137657.1 | CAB70862.2 | |||
BC004365.1 | AAH04365.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048788 | Chr.15:81257750 - 81343926 on Build GRCh38 | Loss | STARD5 IL16 TMC3-AS1 TMC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |