Assay Details
Target Gene Details
Entrez Gene ID: | 50804 |
Gene Name: | myelin expression factor 2 |
Gene Aliases: |
HsT18564, MEF-2, MST156, MSTP156, myEF-2 |
Location: |
Chr.15:48134632-48178517 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832999 | Chr.15:47960097 - 48160208 on Build GRCh38 | Gain | MYEF2 SLC24A5 |
nsv569338 | Chr.15:48104611 - 48160602 on Build GRCh38 | Loss | MYEF2 SLC24A5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |