Assay Details
Target Gene Details
Entrez Gene ID: | 105371031 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.15:89399716-89507395 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371031 |
Target Gene Details
Entrez Gene ID: | 51458 |
Gene Name: | Rh family C glycoprotein |
Gene Aliases: |
C15orf6, PDRC2, RHGK, SLC42A3 |
Location: |
Chr.15:89471407-89496613 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RHCG | NM_001321041.1 | NP_001307970.1 | ||
NM_016321.2 | NP_057405.1 | |||
NR_110261.1 | ||||
AF081497.1 | AAD55748.1 | |||
AF193809.1 | AAF19372.1 | |||
AK290899.1 | ||||
AK298252.1 | ||||
AK298641.1 | ||||
AK301640.1 | ||||
AK310222.1 | ||||
AK313238.1 | ||||
AY257182.1 | AAP81044.1 | |||
BC030965.1 | AAH30965.1 | |||
DC306265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv570386 | Chr.15:89464359 - 89517327 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)