Assay Details
Target Gene Details
Entrez Gene ID: | 321 |
Gene Name: | amyloid beta precursor protein binding family A member 2 |
Gene Aliases: |
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L |
Location: |
Chr.15:28885479-29118315 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | GOLGA8R LOC105370751 NSMCE3 WHAMMP2 ULK4P3 LOC100130111 GOLGA8M DKFZP434L187 GOLGA6L7P CHRFAM7A TJP1 APBA2 LOC101060569 LOC100289656 LOC105370746 FAM189A1 GOLGA8T GOLGA8J PDCD6IPP2 LOC107984752 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | APBA2 LOC100289656 NSMCE3 WHAMMP2 FAM189A1 LOC100130111 GOLGA6L7P PDCD6IPP2 LOC107984752 TJP1 |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | LOC105370751 NSMCE3 ULK4P3 ULK4P2 LOC100130111 GOLGA8H GOLGA6L7P TJP1 LOC100996413 APBA2 FAM189A1 LOC107984752 GOLGA8R WHAMMP2 DKFZP434L187 LOC101927579 CHRFAM7A LOC101060569 LOC100289656 LOC105370746 LOC100288637 GOLGA8T GOLGA8J PDCD6IPP2 ARHGAP11B |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | NSMCE3 WHAMMP2 ULK4P3 LOC100130111 GOLGA6L7P TJP1 APBA2 LOC100289656 FAM189A1 GOLGA8T GOLGA8J PDCD6IPP2 LOC107984752 |
nsv516384 | Chr.15:28920744 - 28996217 on Build GRCh38 | Loss | APBA2 |
nsv1038806 | Chr.15:28933682 - 28965543 on Build GRCh38 | Gain | APBA2 |
nsv568662 | Chr.15:27726493 - 29056123 on Build GRCh38 | Gain | HERC2 HERC2P11 WHAMMP2 HERC2P9 GOLGA8M GOLGA6L7P MIR4509-3 LOC645202 MIR4509-2 OCA2 APBA2 LOC100289656 GOLGA8G GOLGA8F LOC100132202 LOC101929047 PDCD6IPP2 LOC107984752 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |