Assay Details
Target Gene Details
Entrez Gene ID: | 56924 |
Gene Name: | p21 (RAC1) activated kinase 6 |
Gene Aliases: |
PAK5 |
Location: |
Chr.15:40239091-40277487 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAK6 | NM_001276718.1 | NP_001263647.1 | ||
NM_020168.5 | NP_064553.1 | |||
AF276893.1 | AAF82800.1 | |||
AJ236915.1 | CAC18720.1 | |||
AK131522.1 | ||||
DA229537.1 | ||||
HY128655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471240 | Chr.15:40235954 - 40303426 on Build GRCh38 | Loss | PAK6 C15orf56 PLCB2 BUB1B-PAK6 ANKRD63 |
nsv952611 | Chr.15:40237800 - 40284699 on Build GRCh38 | Deletion | PAK6 C15orf56 BUB1B-PAK6 ANKRD63 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |