Assay Details
Target Gene Details
Entrez Gene ID: | 4734 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase |
Gene Aliases: |
NEDD4-1, RPF1 |
Location: |
Chr.15:55826917-55993746 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEDD4 | NM_001284338.1 | NP_001271267.1 | ||
NM_001284339.1 | NP_001271268.1 | |||
NM_001284340.1 | NP_001271269.1 | |||
NM_006154.3 | NP_006145.2 | |||
NM_198400.3 | NP_940682.2 | |||
NR_104302.1 | ||||
XM_011521624.2 | XP_011519926.1 | |||
XM_011521625.2 | XP_011519927.1 | |||
XM_011521626.1 | XP_011519928.1 | |||
XM_011521627.1 | XP_011519929.1 | |||
AK304394.1 | ||||
AK307541.1 | ||||
AL832063.1 | ||||
AL832359.1 | ||||
AL833293.1 | ||||
AL833300.1 | ||||
AY550969.1 | ||||
BC136605.1 | ||||
BC144284.1 | ||||
BC144285.1 | ||||
BC152452.1 | ||||
BC152562.1 | ||||
D42055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833017 | Chr.15:55673759 - 55865714 on Build GRCh38 | Loss | PRTG NEDD4 |
nsv569539 | Chr.15:55430015 - 55963576 on Build GRCh38 | Loss | LOC107984745 PYGO1 DYX1C1-CCPG1 DYX1C1 PRTG NEDD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |