Assay Details
Target Gene Details
Entrez Gene ID: | 1445 |
Gene Name: | c-src tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.15:74782064-74803198 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSK | NM_001127190.1 | NP_001120662.1 | ||
NM_004383.2 | NP_004374.1 | |||
XM_005254165.4 | XP_005254222.1 | |||
XM_017021925.1 | XP_016877414.1 | |||
AK127672.1 | ||||
AK223626.1 | BAD97346.1 | |||
AK290531.1 | ||||
AK298245.1 | ||||
AK310136.1 | ||||
AK312669.1 | ||||
BC104847.1 | AAI04848.1 | |||
BC104875.1 | AAI04876.1 | |||
BC106073.1 | AAI06074.1 | |||
X59932.1 | CAA42556.1 | |||
X60114.1 | CAA42713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss | CYP1A1 CPLX3 EDC3 MIR4513 SCAMP2 CSK MIR6882 ULK3 CYP1A2 LMAN1L |
esv3636909 | Chr.15:74791220 - 74794723 on Build GRCh38 | Loss | CSK |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | SCAMP2 MIR6882 ADPGK-AS1 CCDC33 LOC101929333 LOC732265 STOML1 CYP11A1 GOLGA6C RPP25 C15orf59-AS1 FAM219B ULK3 C15orf39 LOC283731 ISLR UBL7 LOC729739 MIR6881 SCAMP5 CSK CYP1A2 NPTN EDC3 GOLGA6A NPTN-IT1 LOC105376731 LOXL1-AS1 STRA6 PPCDC CYP1A1 HCN4 ISLR2 TBC1D21 SEMA7A UBL7-AS1 CPLX3 CD276 NEO1 ADPGK CLK3 ARID3B COX5A LOC107984728 MPI MIR4513 HIGD2B REC114 C15orf59 BBS4 PML LOXL1 LMAN1L LOC102723750 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |