Assay Details
Target Gene Details
Entrez Gene ID: | 56924 |
Gene Name: | p21 (RAC1) activated kinase 6 |
Gene Aliases: |
PAK5 |
Location: |
Chr.15:40239091-40277487 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAK6 | NM_001276717.1 | NP_001263646.1 | ||
NM_001276718.1 | NP_001263647.1 | |||
NM_020168.5 | NP_064553.1 | |||
AF276893.1 | AAF82800.1 | |||
AJ236915.1 | CAC18720.1 | |||
AK131522.1 | ||||
BC035596.1 | AAH35596.1 | |||
DA191028.1 | ||||
DA229537.1 | ||||
HY128655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471240 | Chr.15:40235954 - 40303426 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3581650 | Chr.15:40254799 - 40256959 on Build GRCh38 | Loss |
![]() ![]() |
nsv952611 | Chr.15:40237800 - 40284699 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)