Assay Details
Target Gene Details
Entrez Gene ID: | 9245 |
Gene Name: | glucosaminyl (N-acetyl) transferase 3, mucin type |
Gene Aliases: |
C2/4GnT, C24GNT, C2GNT2, C2GNTM, GNTM |
Location: |
Chr.15:59611783-59620011 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GCNT3 | NM_004751.2 | NP_004742.1 | ||
BC017032.1 | AAH17032.1 | |||
BP243086.1 | ||||
EF152283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain | MYO1E LDHAL6B BNIP2 CCNB2 GTF2A2 FAM63B MIR2116 FAM81A SLTM RNF111 GCNT3 |
esv3636644 | Chr.15:59550375 - 59641686 on Build GRCh38 | Loss | GTF2A2 GCNT3 |
nsv569605 | Chr.15:59563027 - 59664354 on Build GRCh38 | Loss | BNIP2 GTF2A2 GCNT3 |
esv2668729 | Chr.15:59550283 - 59641656 on Build GRCh38 | Deletion | GTF2A2 GCNT3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |