Assay Details
Target Gene Details
Entrez Gene ID: | 25963 |
Gene Name: | transmembrane protein 87A |
Gene Aliases: |
- |
Location: |
Chr.15:42210452-42273584 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM87A | NM_001286487.1 | NP_001273416.1 | ||
NM_015497.4 | NP_056312.2 | |||
XM_005254287.1 | XP_005254344.1 | |||
XM_006720468.2 | XP_006720531.1 | |||
XM_011521443.2 | XP_011519745.1 | |||
XM_017022067.1 | XP_016877556.1 | |||
AF132733.1 | AAF66444.1 | |||
AK074870.1 | BAC11256.1 | |||
AK075403.1 | BAC11598.1 | |||
AK289591.1 | ||||
AK290150.1 | ||||
AK290484.1 | ||||
AK297816.1 | ||||
AK309915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054102 | Chr.15:42200854 - 42236567 on Build GRCh38 | Gain | TMEM87A VPS39 |
nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss | TMEM87A PLA2G4F VPS39 MIR627 PLA2G4D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |