Assay Details
Target Gene Details
Entrez Gene ID: | 26035 |
Gene Name: | glucuronic acid epimerase |
Gene Aliases: |
HSEPI |
Location: |
Chr.15:69160555-69272209 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLCE | NM_001324091.1 | NP_001311020.1 | ||
NM_001324092.1 | NP_001311021.1 | |||
NM_001324093.1 | NP_001311022.1 | |||
NM_001324094.1 | NP_001311023.1 | |||
NM_015554.2 | NP_056369.1 | |||
XM_005254298.3 | XP_005254355.1 | |||
AY635582.1 | AAT48654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517185 | Chr.15:69137677 - 69260783 on Build GRCh38 | Gain+Loss | GLCE |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |