Assay Details
Target Gene Details
Entrez Gene ID: | 80208 |
Gene Name: | spastic paraplegia 11 (autosomal recessive) |
Gene Aliases: |
ALS5, CMT2X, KIAA1840 |
Location: |
Chr.15:44562696-44663678 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPG11 | NM_001160227.1 | NP_001153699.1 | ||
NM_025137.3 | NP_079413.3 | |||
XM_006720700.1 | XP_006720763.1 | |||
XM_006720701.3 | XP_006720764.1 | |||
XM_017022634.1 | XP_016878123.1 | |||
XM_017022635.1 | 28 | 5202 | XP_016878124.1 | |
XM_017022636.1 | XP_016878125.1 | |||
AB058743.2 | BAB47469.2 | |||
AB470308.1 | ||||
AB470309.1 | ||||
AB470310.1 | ||||
AK091176.1 | BAC03600.1 | |||
BC150640.1 | ||||
BC153879.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |